Skip to Content

D6819

N2-[[5-(Diphenylmethyl)-2-furanyl]carbonyl]-L-Arginine hydrate

≥98% (HPLC), powder

Synonym(s):

Compound 5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H26N4O4 · xH2O
CAS Number:
Molecular Weight:
434.49 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.NC(=N)NCCC[C@H](NC(=O)c1ccc(o1)C(c2ccccc2)c3ccccc3)C(O)=O

InChI

1S/C24H26N4O4.H2O/c25-24(26)27-15-7-12-18(23(30)31)28-22(29)20-14-13-19(32-20)21(16-8-3-1-4-9-16)17-10-5-2-6-11-17;/h1-6,8-11,13-14,18,21H,7,12,15H2,(H,28,29)(H,30,31)(H4,25,26,27);1H2/t18-;/m0./s1

InChI key

SFOGIFDRPRBBFF-FERBBOLQSA-N

Biochem/physiol Actions

N2-[[5-(Diphenylmethyl)-2-furanyl]carbonyl]-L-Arginine hydrate is a C3a receptor antagonist.
N2-[[5-(Diphenylmethyl)-2-furanyl]carbonyl]-L-Arginine hydrate is a C3a receptor antagonist. The Complement component 3a receptor is a G protein-coupled receptor protein involved in the complement system.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library