Skip to Content
Merck

D6777

1,3:4,6-Di-O-benzylidene-D-mannitol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
1627753
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C

SMILES string

O[C@@H]1COC(O[C@H]1[C@@H]2OC(OC[C@H]2O)c3ccccc3)c4ccccc4

InChI

1S/C20H22O6/c21-15-11-23-19(13-7-3-1-4-8-13)25-17(15)18-16(22)12-24-20(26-18)14-9-5-2-6-10-14/h1-10,15-22H,11-12H2/t15-,16-,17-,18-,19?,20?/m1/s1

InChI key

NVEGGHPETXMRSV-NIJYPJQDSA-N

Application

Stable blocked sugar for chiral synthesis and asymmetric reduction.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library