Skip to Content
Merck

D6418

DEAE-Cellulose

Fast Flow, fibers

Synonym(s):

Cellulose DEAE, Diethylaminoethyl-cellulose

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
UNSPSC Code:
47101511
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

DEAE-Cellulose, Fast Flow, fibers

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

SMILES string

OC[C@@H]1O[C@@H](O[C@H]2[C@@H](O)[C@H](O)[C@@H](O)O[C@H]2CO)[C@@H](O)[C@H](O)[C@H]1O

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

form

fibers

Looking for similar products? Visit Product Comparison Guide

Application

DEAE-Cellulose is used in ion exchange chromatography, protein chromatography and anion exchange media. DEAE cellulose has been used to study acute myocardial infarction.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Martin-Sanchez et al.
Experimental parasitology, 92(1), 57-63 (1999-05-18)
Isoenzyme electrophoresis and RAPD techniques were used to study the genetic polymorphism of different Spanish isolates of Theileria annulata in the schizont and piroplasm stages. Enzyme activity attributable to the parasite was detected in only 5 of the 13 loci
Jana Stránská et al.
Biochimie, 89(1), 135-144 (2006-09-23)
A series of N,N'-bis(2-pyridinylmethyl)diamines was synthesized and characterized for their inhibition effects towards plant copper-containing amine oxidase (EC 1.4.3.6) and polyamine oxidase (EC 1.5.3.11), which mediate the catabolic regulation of cellular polyamines. Even though these enzymes catalyze related reactions and
Buuan Lam et al.
Analytical chemistry, 78(24), 8194-8199 (2006-12-15)
A passive sampler for the isolation of dissolved organic matter (DOM) from freshwater environments is described. The sampler consists of a molecular weight selective membrane (1000 kDa) and an anion exchange resin (diethylaminoethylcellulose (DEAE-cellulose)). NMR indicates the samplers isolate DOM
Collagen-Induced Arthritis in Rats
Griffiths, M.M., et al.
Arthritis Research, 136(II), 201-214 (2007)
Xiang Ding et al.
International journal of biological macromolecules, 50(3), 613-618 (2012-01-31)
In this study, a novel heteropolysaccharide was isolated from the fruiting bodies of Boletus speciosus Forst through DEAE-cellulose column and Sephadex G-200 column. The Boletus speciosus Forst polysaccharide (BSFP-1) had a molecular weight of 1.33×10(4) Da and was mainly composed

Related Content

Product Information Sheet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service