Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C15H8Cl2O3
CAS Number:
Molecular Weight:
307.13
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Skip To
InChI key
QMSGNLMBWFRALO-UHFFFAOYSA-N
SMILES string
Oc1ccc2C(=O)C(=COc2c1)c3ccc(Cl)cc3Cl
InChI
1S/C15H8Cl2O3/c16-8-1-3-10(13(17)5-8)12-7-20-14-6-9(18)2-4-11(14)15(12)19/h1-7,18H
assay
≥98% (HPLC)
form
solid
color
yellow
solubility
DMSO: >20 mg/mL
Biochem/physiol Actions
DCHC is a SIRT1 activator. It increases peroxisome proliferator-activated receptor gamma co-activator 1α (PGC1α) expression, resulting in mitochondrial biogenesis.
DCHC is a SIRT1 activator. It increases peroxisome proliferator-activated receptor gamma co-activator 1α (PGC1α) expression, resulting in mitochondrial biogenesis. DCHC activates SIRT1 at low micromolar concentrations (5–10 μM) and increases SIRT1 activity at 100 μM. It is different from 5,7,4′-T in that it induces the activation of recombinant SIRT1, but does not increase SIRT1 expression.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hsiu-Chung Ou et al.
Molecular nutrition & food research, 63(10), e1801353-e1801353 (2019-03-21)
Atherosclerotic cardiovascular disease is the most prevalent cause of mortality and morbidity. Fucoxanthin (FX) possesses anti-hypertensive and anti-obesity properties. However, the molecular mechanisms underlying the inhibitory effects of FX on oxidized low-density lipoprotein (oxLDL)-induced oxidative injuries in human endothelial cells
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service