Skip to Content
Merck

D5419

n-Decyl α-D-glucopyranoside

~98% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H32O6
CAS Number:
Molecular Weight:
320.42
UNSPSC Code:
12352200
EC Number:
249-846-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

assay

~98% (GC)

storage temp.

−20°C

SMILES string

CCCCCCCCCCOC1OC(CO)C(O)C(O)C1O



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Brown, G.M., et al.
Canadian Journal of Chemistry, 48, 2525-2525 (1970)