Skip to Content

D5399

3,5-Dibenzyloxyacetophenone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H5CH2O)2C6H3COCH3
CAS Number:
Molecular Weight:
332.39
EC Number:
249-315-7
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

60-62 °C (lit.)

SMILES string

CC(=O)c1cc(OCc2ccccc2)cc(OCc3ccccc3)c1

InChI

1S/C22H20O3/c1-17(23)20-12-21(24-15-18-8-4-2-5-9-18)14-22(13-20)25-16-19-10-6-3-7-11-19/h2-14H,15-16H2,1H3

InChI key

KOJXGMJOTRYLBD-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library