Skip to Content
Merck

D4790

N,N-Dimethyl-p-phenylenediamine sulfate salt

Synonym(s):

4-(Dimethylamino)aniline sulfate salt, 4-Amino-N,N-dimethylaniline sulfate salt, DMPPDA

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
(CH3)2NC6H4NH2·H2SO4
CAS Number:
Molecular Weight:
234.27
EC Number:
208-636-2
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
4612278
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

495 °C (lit.)

mp

200-205 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.H2O4S/c1-10(2)8-5-3-7(9)4-6-8;1-5(2,3)4/h3-6H,9H2,1-2H3;(H2,1,2,3,4)

InChI key

GLUKPDKNLKRLHX-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudineia R Silva et al.
Talanta, 85(3), 1703-1705 (2011-08-03)
A spectrophotometric flow injection procedure involving N,N-dimethyl-p-phenylenediamine (DMPD) is applied to the sulfide monitoring of a sugar fermentation by Saccharomyces cerevisiae under laboratory conditions. The gaseous chemical species evolving from the fermentative process, mainly CO(2), are trapped allowing a cleaned
F J Alcaín et al.
Redox report : communications in free radical research, 3(5-6), 287-293 (1998-12-16)
Swiss 3T3 fibroblasts can be weakly stimulated to grow by bombesin, epidermal growth factor or ceruloplasmin when cells are maintained in Dulbecco's Modified Essential Medium (DMEM), the pH of which is 7.75. Addition of insulin synergizes with the other mitogens.
P K Wong et al.
Journal of applied microbiology, 85(1), 79-87 (1998-08-29)
Klebsiella pneumoniae RS-13 and Acetobacter liquefaciens S-1, both methyl red (MR)-degrading bacterial strains, degraded N,N'-dimethyl-p-phenylenediamine (DMPD) under aerobic conditions. DMPD, a toxic and mutagenic aromatic amine, is formed during the reductive cleavage of azo dyes such as MR. The effects
C Li et al.
The Biochemical journal, 300 ( Pt 1), 31-36 (1994-05-15)
C-terminal alpha-amidation is a structural feature essential to the biological activity of many peptide hormones. Peptidylglycine alpha-amidating mono-oxygenase (PAM; EC 1.14.17.3) catalyses conversion of glycine-extended peptide hormone precursors into their corresponding alpha-hydroxyglycine derivatives. This reaction is the first step in
C Störle et al.
Chemico-biological interactions, 83(3), 271-291 (1992-08-28)
N,N-Dimethyl-p-phenylenediamine (DMPD) reacted directly with oxyhemoglobin under formation of ferrihemoglobin and, presumably, the N,N-dimethyl-p-phenylenediamine radical cation (DMPP.+). The apparent second-order rate constant of this reaction was 1 M-1 s-1 (pH 7.4, 37 degrees C). The reaction rate was diminished by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service