Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H21N3O2
CAS Number:
Molecular Weight:
311.38
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality Segment
assay
≥98% (HPLC)
form
solid
solubility
DMSO: 22 mg/mL
storage temp.
2-8°C
SMILES string
CCN(CC)c1ccc(cc1)\C=N\NC(=O)c2ccc(O)cc2
InChI
1S/C18H21N3O2/c1-3-21(4-2)16-9-5-14(6-10-16)13-19-20-18(23)15-7-11-17(22)12-8-15/h5-13,22H,3-4H2,1-2H3,(H,20,23)/b19-13+
InChI key
WLKOCYWYAWBGKY-CPNJWEJPSA-N
Biochem/physiol Actions
DY 131 is an estrogen related receptor ERRβ and ERRy agonist with minimal activity at ERRα, ERα and ERβ
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offers many products related to Nuclear Receptors (Steroids) for your research needs.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D3946-25MG | 04061832867076 |
| D3946-5MG | 04061832867083 |
