Skip to Content
Merck

Skip To

D3254

7,12-Dimethylbenz[a]anthracene

≥95%

Synonym(s):

1,4-Dimethyl-2,3-benzophenanthrene, 9,10-Dimethyl-1,2-benzanthracene, DMBA

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H16
CAS Number:
Molecular Weight:
256.34
UNSPSC Code:
12352200
NACRES:
NA.25
PubChem Substance ID:
EC Number:
200-359-5
Beilstein/REAXYS Number:
1912135
MDL number:

assay

≥95%

form

powder

mp

122-123 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

Cc1c2ccccc2c(C)c3c1ccc4ccccc34

InChI

1S/C20H16/c1-13-16-8-5-6-9-17(16)14(2)20-18(13)12-11-15-7-3-4-10-19(15)20/h3-12H,1-2H3

InChI key

ARSRBNBHOADGJU-UHFFFAOYSA-N

General description

7,12-Dimethylbenz[a]anthracene belongs to the category of polycyclic aromatic hydrocarbons.[1] It is a carcinogenic organic pollutant generated from the incomplete combustion of gasoline and coal.

Application

7,12-Dimethylbenz[a]anthracene was used:
  • to study the effects of Cimicifuga racemosa (CR) extract on the growth of mammary tumor induced by 7,12-Dimethylbenz[a]anthracene[2]
  • in the study to compare the anti-carcinogenic properties of four red wine polyphenols as an initiator to cause skin cancer in CD-1 mouse model[3]
  • to induce epithelial carcinogenicity in order to study the apoptotic, proliferating and p12doc-1 profiles of normal, hyperplastic, dysplastic and malignant oral epithelium in the cheek pouch of the Syrian hamster[4]
  • to cause mammary tumors to examine the causes and prevention of triacylglycerol accumulation in rat liver due to tamoxifen[5]

Biochem/physiol Actions

7,12-Dimethylbenz[a]anthracene has been found to be cytotoxic in nature, having carcinogenic properties. It also shows selective necrosis and hemorrhaging in the adrenal gland along with an inhibitory effect on plasma alkaline phosphatase levels. Its damaging effects are due to its ability to form charge transfer complexes by donating electrons to appropriate electron acceptors.[4][1]

pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steroid receptor coactivators, HER-2 and HER-3 expression is stimulated by tamoxifen treatment in DMBA-induced breast cancer
Moi LLH, et al.
BMC Cancer, 12(1), 247-247 (2012)
A comparison of the anticarcinogenic properties of four red wine polyphenols
Soleas GJ, et al.
Clinical Biochemistry, 35(2), 119-124 (2002)
T Ishikawa et al.
Anticancer research, 19(5A), 3749-3752 (2000-01-08)
A two-stage mouse skin carcinogenesis model was used to examine the effects of IP6 on initiation and promotion phases of tumorigenesis. Seven week old ICR female mice were divided into 6 groups, each consisting of 20 animals. Initiation was performed
Yohko Kohno et al.
Oral oncology, 38(3), 274-280 (2002-04-30)
Disruption of the homeostatic balance between proliferation and apoptosis is widely believed to contribute to human oral carcinogenesis. Using the Syrian hamster oral cancer model, we examined normal, hyperplastic, dysplastic and malignant oral epithelium for the fraction of apoptotic, proliferating
Regio-and stereoselective metabolism of 7, 12-dimethylbenz [a] anthracene by Mycobacterium vanbaalenii PYR-1
Moody JD, et al.
Applied and Environmental Microbiology, 69(7), 3924-3931 (2003)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Global Trade Item Number

SKUGTIN
D3254-1G04061835326808
D3254-5G04061833563052
D3254-100MG04061835326792

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service