Skip to Content
Merck

D2380

DNP-L-valine

Synonym(s):

N-(2,4-Dinitrophenyl)-L-valine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C11H13N3O6
CAS Number:
Molecular Weight:
283.24
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
216-909-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

storage temp.

−20°C

SMILES string

CC(C)C(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service