Skip to Content
Merck

D1375

Dansyl-L-leucine cyclohexylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C18H24N2O4S · C6H13N
CAS Number:
Molecular Weight:
463.63
UNSPSC Code:
12352200
PubChem Substance ID:
EC Number:
256-018-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

KMKWMTLYBLHMGU-RSAXXLAASA-N

InChI

1S/C18H24N2O4S.C6H13N/c1-12(2)11-15(18(21)22)19-25(23,24)17-10-6-7-13-14(17)8-5-9-16(13)20(3)4;7-6-4-2-1-3-5-6/h5-10,12,15,19H,11H2,1-4H3,(H,21,22);6H,1-5,7H2/t15-;/m0./s1

SMILES string

NC1CCCCC1.CC(C)C[C@H](NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O

storage temp.

−20°C

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service