Skip to Content

D1018

2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose

Synonym(s):

D-Fructopyranose diacetonide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

CC1(C)O[C@@H]2CO[C@@]3(CO)OC(C)(C)O[C@H]3[C@@H]2O1

InChI

1S/C12H20O6/c1-10(2)15-7-5-14-12(6-13)9(8(7)16-10)17-11(3,4)18-12/h7-9,13H,5-6H2,1-4H3/t7-,8-,9+,12+/m1/s1

InChI key

PSSHGMIAIUYOJF-XBWDGYHZSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library