Skip to Content
Merck

D0914

Dideoxycytidine-3H

buffered aqueous solution

Synonym(s):

2′,3′-Dideoxycytidine-2′,3′-3H, ddC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12142207
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

extent of labeling

40-60 Ci/mmol

storage temp.

2-8°C

SMILES string

[H]C1C([H])C(OC1CO)N2C=CC(N)=NC2=O

Biochem/physiol Actions

Tritiated (3H) dideoxycytidine is supplied ready-for-use in a double antibody radioimmunoassay. Contains less than 1 μCi of 3H-ddC in 10.5 ml.

Physical form

Solution in 0.1 M phosphate buffer, pH 7.4, containing 200 μg/ml rabbit IgG, protein stabilizers and 15 mM sodium azide.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content