Skip to Content
Merck

D0569

DT-3 trifluoroacetate salt

≥95% (HPLC), lyophilized powder

Synonym(s):

Drosophila Antennapedia homeodomain (43-58)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C152H259N53O27S · xC2HF3O2
CAS Number:
Molecular Weight:
3293.09 (free base basis)
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

lyophilized powder

composition

Peptide content, ~64%

color

white

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc6c[nH]cn6)C(N)=O

Biochem/physiol Actions

Cell permeable cGMP-dependent protein kinase type Ia (PKG) inhibitor


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Thomas Krieg et al.
American journal of physiology. Heart and circulatory physiology, 288(4), H1976-H1981 (2004-12-14)
Bradykinin and acetylcholine (ACh) trigger preconditioning by ATP-sensitive K(+) (K(ATP)) channel-dependent production of reactive oxygen species (ROS). Recent evidence suggests that ROS production may in turn be influenced by cGMP-dependent protein kinase (PKG). This study utilized DT-2 and DT-3 peptides
Wolfgang R G Dostmann et al.
Pharmacology & therapeutics, 93(2-3), 203-215 (2002-08-23)
The structural similarity of cyclic GMP-dependent protein kinase (cGPK) and cyclic AMP-dependent protein kinase (cAPK) has made it difficult to study cGPK pathways independent of those mediated by cAPK, primarily due to the lack of potent and selective cGPK inhibitors.
Highly specific, membrane-permeant peptide blockers of cGMP-dependent protein kinase Ia inhibit NO-induced cerebral dilation
Dostmann, Wolfgang R.G. et al.
Proceedings of the National Academy of Sciences of the USA, 19, 14772 - 14777 (2000)