Skip to Content
Merck

D0300

Difucosyllacto-N-hexaose A from human milk

≥90% (HPLC)

Synonym(s):

DFLNH(a), DFLN(a)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C52H88N2O39
CAS Number:
Molecular Weight:
1365.25
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

CC1OC(OC2C(NC(C)=O)C(OC(CO)C2OC3OC(CO)C(O)C(O)C3O)OC4C(O)C(CO)OC(OC5C(CO)OC(OC6C(O)C(CO)OC(OC(C(O)CO)C(O)C(O)C=O)C6O)C(NC(C)=O)C5OC7OC(C)C(O)C(O)C7O)C4O)C(O)C(O)C1O

Other Notes

Octasaccharide containing the Lewis X substructure




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library