Skip to Content
Merck

D0126

2′-Deoxyinosine 5′-monophosphate sodium salt

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H13N4O7P · xNa+
CAS Number:
Molecular Weight:
332.21 (free acid basis)
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
239-090-3
Assay:
≥97% (HPLC)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless to faintly yellow
Storage temp.:
−20°C

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥97% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na].OC1CC(OC1COP(O)(O)=O)n2cnc3C(=O)N=CNc23

InChI

1S/C10H13N4O7P.2Na/c15-5-1-7(21-6(5)2-20-22(17,18)19)14-4-13-8-9(14)11-3-12-10(8)16;;/h3-7,15H,1-2H2,(H,11,12,16)(H2,17,18,19);;/q;2*+1/p-2/t5-,6+,7+;;/m0../s1

InChI key

VDPLPYMIJCLJEF-OJSHLMAWSA-L

Application

2′-Deoxyinosine 5′-monophosphate (dIMP) is generated in DNA upon the deamination of adenosine. 2′-Deoxyinosine 5′-monophosphate may be used to study nitrosative deamination processes.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
D0126-5MG04061833006702

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service