Skip to Content
Merck

D004

R(−)-Apomorphine hydrochloride

small crystals

Synonym(s):

R(–)-10,11-Dihydroxyaporphine, R(–)-APO

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C17H17NO2 · HCl
CAS Number:
Molecular Weight:
303.78
UNSPSC Code:
41116107
EC Number:
206-243-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

small crystals

optical activity

[α]22/D −75.75°, c = 0.37 in methanol(lit.)

solubility

alcohol: 20 mg/mL, 0.15 M NaCl: insoluble, H2O: soluble (Dissolve in oxygen-free boiled water containing 0.1% sodium metabisulfite or other antioxidant.)

storage temp.

2-8°C

Biochem/physiol Actions

Dopamine receptor agonist.

Disclaimer

Hygroscopic, photosensitive

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Wilcox et al.
Journal of pharmaceutical sciences, 69(8), 974-976 (1980-08-01)
Ascorbic acid (100 mg/ml) and sodium bisulfite (0.5 and 20 mg/ml) prevented more than 10% oxidation of apomorphine hydrochloride in water maintained at room temperature over 1-3 days. Refrigeration at 5 degrees prevented oxidation of apomorphine hydrochloride in aqueous solutions
Attila Sipos et al.
Bioorganic & medicinal chemistry, 16(7), 3773-3779 (2008-02-22)
A novel synthesis has been elaborated for the pharmacologically remarkable 2-arylapomorphines described and characterized in the last few years. This new procedure contains two alternative synthetic routes and has allowed the preparation of several hitherto unknown compounds as well. The
Gessa, et al., eds.
Apomorphine and Other Dopaminomimetics, 1, 2 (1981)
K Ishige et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 21(16), 6069-6076 (2001-08-07)
Oxidative stress is thought to be the cause of nerve cell death in many CNS pathologies, including ischemia, trauma, and neurodegenerative disease. Glutamate kills nerve cells that lack ionotropic glutamate receptors via the inhibition of the cystine-glutamate antiporter x(c)(-), resulting
N S Kula et al.
Life sciences, 37(11), 1051-1057 (1985-09-16)
The optical isomers of apomorphine (APO) and N-propylnorapomorphine (NPA) were interacted with three biochemical indices of dopamine (DA) receptors in extrapyramidal and limbic preparations of rat brain tissue. There were consistent isomeric preferences for the R(-) configuration of both DA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service