CLS4366
Corning® universal rack for 15 mL and 50 mL conical bottom centrifuge tubes
Synonym(s):
centrifuge tube racks, tube racks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polystyrene
sterility
non-sterile
feature
Holds 25 x 15 mL tubes
Holds 25 x 50 mL tubes
packaging
case of 20
pack of 20
manufacturer/tradename
Corning 4366
capacity
25 tubes
color
white
Related Categories
General description
Corning® Universal Rack for 15 mL and 50 mL Conical Bottom Centrifuge Tubes
- Manufactured out of polystyrene foam
- Economical storage of 15 mL and 50 mL centrifuge tubes
- Designed to hold up to 25 x 15 mL or 50 mL conical bottom centrifuge tubes
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service