CLS3775
Corning® CrystalEX™ 384 well protein crystallization plates
cyclic olefin copolymer (COC), flat bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well protein crystallization plates, protein crystallization plates
Select a Size
CZK 42,200.00
Select a Size
About This Item
cyclic olefin copolymer (COC)
flat bottom
CZK 42,200.00
Recommended Products
material
clear cyclic olefin copolymer (COC) plate
cyclic olefin copolymer (COC)
flat bottom
sterility
non-sterile
feature
lid: no
plate format: 384 well standard (1 protein well/reservoir well)
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3775
reservoir volume
105 μL
size
384 wells
well volume
4 μL , protein well volume
suitability
suitable for (cell cuture)
Related Categories
General description
- 384 well microplate designed for full automation in crystal screening, built to meet industry standards for 384 well microplate footprint and well locations
- Features 192 reservoir wells (105 μL/well) and 192 corresponding protein wells (4 μL/well)
- Reservoir wells and protein wells are positioned to be compatible with multihead dispensing equipment (up to 96 well heads)
- Manufactured from an advanced polymer with high resistance to commonly used solvents, including acetone, acetic acid, butanone, ethanol, isopropanol, methanol, DMSO, nitric acid (65%), sulfuric acid (40%), hydrochloric acid (36%), and ammonia solution (33%)
- Low background polarization and high optical clarity allows protein crystals to be easily viewed under polarized light with minimal background interference
- Low water absorption of the polymer prevents loss of protein drop volume
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)