Skip to Content
Merck

Skip To

CLS3729BC

Corning® 1536 well plate, with generic barcode

size 1536 wells, NBS surface, flat bottom white polystyrene wells

Synonym(s):

1536-well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 ea
Please contact Customer Service for Availability
62 700,00 CZK

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
flat bottom white polystyrene wells, polystyrene plate
Size:
1536 wells

62 700,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom white polystyrene wells, polystyrene plate

sterility

non-sterile

feature

lid: no, skirt, NBS surface

packaging

pack of 10, case of 50 ea, case of 50

manufacturer/tradename

Corning 3729BC

maximum volume

12.5 μL

size

1536 wells

working volume

8 μL

color

white

suitability

suitable for (cell cuture)

binding type

NBS™ (non-binding surface), low binding surface

General description

Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")
Generic barcoded Corning 1536 well microplates are now available. Code 128 bar code format ensures compatibility with most bar code scanning and software systems. To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code.

  • Resistant to temperature ranges of -80ºC to +52ºC
  • 2.8 mm distance between well bottoms to plate bottom
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface

Microplate Features
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness -minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
  • Working volume of up to 10 μL


Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS3727BCCLS3893BCCLS3728BC
description

size 1536 wells, NBS surface, flat bottom white polystyrene wells

description

size 1536 wells, flat bottom white polystyrene wells, sterile

description

size 1536 wells, flat bottom clear polystyrene wells, black plate

description

size 1536 wells, flat bottom wells black polystyrene wells, NBS surface, 50/cs, non-sterile

size

1536 wells

size

1536 wells

size

1536 wells

size

1536 wells

suitability

suitable for (cell cuture)

suitability

suitable for (cell cuture)

suitability

suitable for (cell cuture)

suitability

suitable for (cell cuture)

material

polystyrene plate, flat bottom white polystyrene wells

material

flat bottom white polystyrene wells, polystyrene plate

material

polystyrene plate, flat bottom clear polystyrene wells

material

polystyrene plate, flat bottom wells black polystyrene wells

manufacturer/tradename

Corning 3729BC

manufacturer/tradename

Corning 3727BC

manufacturer/tradename

Corning 3893BC

manufacturer/tradename

Corning 3728BC

packaging

case of 50 ea, pack of 10

packaging

pack of 10, case of 50, case of 50 ea

packaging

case of 50 ea, pack of 10

packaging

pack of 10, case of 50 ea

feature

NBS surface, lid: no, skirt

feature

skirt, lid

feature

lid

feature

lid: no, skirt


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library