Skip to Content
Merck

Skip To

CLS3261

Corning® Ultra-low attachment culture dishes

60mm petri dish, low-attachment

Synonym(s):

cell culture flasks, tissue culture flasks

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 ea
Please contact Customer Service for Availability
20 000,00 CZK

About This Item

NACRES:
NB.13
UNSPSC Code:
41121812
Material:
polystyrene , round
Sterility:
sterile
Manufacturer/tradename:
Corning 3261
Packaging:
case of 20

20 000,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene , round

sterility

sterile

packaging

case of 20

manufacturer/tradename

Corning 3261

D × H

60 mm × 15 mm

surface area

21 cm2

color

clear

General description

  • Ultra Low Attachment dishes feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Sterilized by gamma irradiation and certified nonpyrogenic
  • Actual inside growth surface diameter is 51.4mm
  • Height is 15mm
  • Manufactured from optically-clear virgin polystyrene
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS430588CLS430589CLS430591
description

60mm petri dish, low-attachment

description

D × H 35 mm × 10 mm

description

D × H 60 mm × 15 mm

description

D × H 100 mm × 20 mm

manufacturer/tradename

Corning 3261

manufacturer/tradename

Corning 430588

manufacturer/tradename

Corning 430589

manufacturer/tradename

Corning 430591

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

material

round , polystyrene

material

polystyrene , round

material

round , polystyrene

material

polystyrene , round

packaging

case of 20

packaging

sleeve of 20

packaging

sleeve of 20

packaging

sleeve of 20

surface area

21 cm2

surface area

8 cm2

surface area

21 cm2

surface area

55 cm2

D × H

60 mm × 15 mm

D × H

35 mm × 10 mm

D × H

60 mm × 15 mm

D × H

100 mm × 20 mm


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library