Skip to Content
Merck

Skip To

CLS2017

Corning® cap inserts for cryogenic vials

red

Synonym(s):

Corning cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 ea
Please contact Customer Service for Availability
1 450,00 CZK

About This Item

NACRES:
NB.13
UNSPSC Code:
41121704
Color:
red
Sterility:
non-sterile

1 450,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

manufacturer/tradename

Corning 2017

color

red

General description

  • Cap inserts provide color coding for easy sample identification
  • Inserts are packaged in resealable bags
  • Cap inserts fit all Corning cryogenic vials except Corning No. 430289

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS2016CLS2018CLS2019
color

red

color

blue

color

green

color

yellow

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

manufacturer/tradename

Corning 2017

manufacturer/tradename

Corning 2016

manufacturer/tradename

Corning 2018

manufacturer/tradename

Corning 2019


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library