Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NA.26
UNSPSC Code:
12352200
shipped in
wet ice
storage temp.
−20°C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Z Szelényi
Peptides, 22(8), 1245-1250 (2001-07-18)
Thermoregulatory effects of cholecystokinin (CCK) peptides are reviewed with special emphasis on two types of responses, that is hypothermia or hyperthermia. In rodents exposed to cold a dose-dependent hypothermia has been observed on peripheral injection of CCK probably acting on
Ming Jiang et al.
Journal of pain research, 12, 2009-2026 (2019-07-17)
Preoperative anxiety is associated with postoperative hyperalgesia; however, few studies have investigated the mechanism underlying this association in female surgical patients. Research has suggested that ON cells in the rostral ventromedial medulla (RVM) receive nerve impulses via cholecystokinin 2 (CCK2)
Olivier Roca-Lapirot et al.
Pain, 160(2), 345-357 (2018-10-04)
Pain is associated with negative emotions such as anxiety, but the underlying neurocircuitry and modulators of the association of pain and anxiety remain unclear. The neuropeptide cholecystokinin (CCK) has both pronociceptive and anxiogenic properties, so we explored the role of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service