Reactivity: Anti-CD44 v6, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93070778
The antibody is directed against the variant region of human CD44 encoded by exon 6. The antibody is suitable for immunohistology, but if used for FACS staining binding must be performed at room temperature.
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)