Skip to Content
Merck
All Photos(1)

Documents

NCI-H292

91091815, human lung, Epithelial

Synonym(s):

H292 Cells, NCI-HUT-292 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H292, 91091815

biological source

human lung

growth mode

Adherent

karyotype

Modal no. 47

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human lung, mucoepidermoid carcinoma

Cell Line Description

Derived from a cervical node metastasis of a pulmonary mucoepidermoid carcinoma from a 32 year old female. Cells retain mucoepidermoid characteristics in culture and support replication of Hepatitis B virus. Some of the cells contain numerous small mucin-containing granules and are L-DOPA decaroxylase negative. This cell line has been selected as a prototype for transfecting subgenome fragments into human cells for studying the role of HBV and its individual genes in the pathogenesis of viral hepatitis and liver cancer. The cells are mucicarmine positive and also stain positively for keratin and vimentin but negative for neurofilament triplet protein. Ethnicity: Black.

Application

NCI-H292 has been used to determine the ability of the cell line to support the replication of mumps virus.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11,12
D16S539: 9,13
D5S818: 13
D7S820: 10
THO1: 8
TPOX: 8,11
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service