Skip to Content
Merck
All Photos(1)

Key Documents

Y13-238

89111411, Reactivity: Anti-p21 Protein, Harvey Murine Sarcoma Virus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Rat Hybridoma

ImmunoDon

Osborne-Mendel rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-p21 Protein, Harvey Murine Sarcoma Virus

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat Osborne-Mendel spleen

Cell Line Description

The antibody is directed against the 21kDa transforming protein (p21) encoded by the v-ras gene of Harvey Murine Sarcoma Virus. It also reacts with the p21 encoded by BALB-MuSV and the 29kDa gag-ras fusion protein encoded by Ra-SV.

Immunogen

Ha-MuSV transformed syngeneic NRK cells

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells will semi-attach. Shake cells off or use trypsin to remove cells from flask

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service