Skip to Content
Merck

C9455

Cellobiitol

≥98%

Synonym(s):

4-O-β-D-Glucopyranosyl-D-glucitol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98%

SMILES string

OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library