Skip to Content
Merck

C8662

C-Peptide Fragment 3-33 human

≥85% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C129H211N35O48
CAS Number:
Molecular Weight:
3020.26
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥85% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)CCC(O)=O)C(C)C)C(C)C)C(=O)NCC(=O)NCC(=O)NCC(=O)N1CCC[C@H]1C(=O)NCC(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C129H211N35O48/c1-58(2)42-77(155-113(195)74(31-39-102(184)185)151-127(209)105(65(15)16)162-114(196)71(25-32-88(131)167)145-95(174)54-141-126(208)104(64(13)14)161-115(197)72(26-33-89(132)168)150-118(200)80(45-61(7)8)156-121(203)83(48-103(186)187)159-112(194)73(30-38-101(182)183)148-107(189)67(18)143-109(191)69(130)24-36-99(178)179)111(193)138-50-93(172)135-49-92(171)136-55-98(177)163-40-20-22-86(163)124(206)140-51-94(173)142-66(17)106(188)137-52-96(175)146-84(56-165)122(204)157-81(46-62(9)10)119(201)152-75(27-34-90(133)169)128(210)164-41-21-23-87(164)125(207)160-78(43-59(3)4)116(198)144-68(19)108(190)154-79(44-60(5)6)117(199)149-70(29-37-100(180)181)110(192)139-53-97(176)147-85(57-166)123(205)158-82(47-63(11)12)120(202)153-76(129(211)212)28-35-91(134)170/h58-87,104-105,165-166H,20-57,130H2,1-19H3,(H2,131,167)(H2,132,168)(H2,133,169)(H2,134,170)(H,135,172)(H,136,171)(H,137,188)(H,138,193)(H,139,192)(H,140,206)(H,141,208)(H,142,173)(H,143,191)(H,144,198)(H,145,174)(H,146,175)(H,147,176)(H,148,189)(H,149,199)(H,150,200)(H,151,209)(H,152,201)(H,153,202)(H,154,190)(H,155,195)(H,156,203)(H,157,204)(H,158,205)(H,159,194)(H,160,207)(H,161,197)(H,162,196)(H,178,179)(H,180,181)(H,182,183)(H,184,185)(H,186,187)(H,211,212)/t66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,104-,105-/m0/s1

InChI key

VOUAQYXWVJDEQY-QENPJCQMSA-N

Gene Information

human ... INS(3630)



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Igano, K
Bulletin of the Chemical Society of Japan, 54, 3088-3088 (1981)
H P Vogt et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 357(1), 107-116 (1976-01-01)
Statistical analysis of protein sequences lends itself to the identification of regions with a definite inclination to adopt specific main-chain conformations. Application of the model of Chou and Fasman[1,2] to porcine proinsulin C-peptide localizes the tendency to form a helix