Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CeCl3 · 7H2O
CAS Number:
Molecular Weight:
372.58
UNSPSC Code:
12352200
MDL number:
density
~3.94 g/mL at 25 °C (lit.)
SMILES string
[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ce](Cl)Cl
InChI
1S/Ce.3ClH.7H2O/h;3*1H;7*1H2/q+3;;;;;;;;;;/p-3
InChI key
KPZSTOVTJYRDIO-UHFFFAOYSA-K
Still not finding the right product?
Explore all of our products under Cerium(III) chloride heptahydrate
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.