C7934
2-Chloro-N6-cyclopentyl-1-deazaadenosine
Synonym(s):
DCCA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H21ClN4O4
CAS Number:
Molecular Weight:
368.82
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
SMILES string
OCC1OC(C(O)C1O)n2cnc3c(NC4CCCC4)cc(Cl)nc23
InChI
1S/C16H21ClN4O4/c17-11-5-9(19-8-3-1-2-4-8)12-15(20-11)21(7-18-12)16-14(24)13(23)10(6-22)25-16/h5,7-8,10,13-14,16,22-24H,1-4,6H2,(H,19,20)
InChI key
ZQSTWOIIMKNFPE-UHFFFAOYSA-N
Biochem/physiol Actions
A selective A1 adenosine receptor agonist
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bin Yu et al.
BMC genomics, 19(1), 478-478 (2018-06-20)
Apoptosis is associated with some human diseases, including cancer, autoimmune disease, neurodegenerative disease and ischemic damage, etc. Apoptosis proteins subcellular localization information is very important for understanding the mechanism of programmed cell death and the development of drugs. Therefore, the
Nam D Nguyen et al.
PLoS computational biology, 16(4), e1007677-e1007677 (2020-04-03)
The molecular mechanisms and functions in complex biological systems currently remain elusive. Recent high-throughput techniques, such as next-generation sequencing, have generated a wide variety of multiomics datasets that enable the identification of biological functions and mechanisms via multiple facets. However
C Cano et al.
The Journal of pharmacology and experimental therapeutics, 261(2), 660-668 (1992-05-01)
The subtype of adenosine receptor linked to cardiac prostacyclin (PGI2) synthesis, measured as immunoreactive 6-keto-PGF1 alpha, was investigated in the rabbit heart perfused with Krebs' buffer at 20 ml/min. Adenosine (6.4-50 nmol) decreased 6-keto-PGF1 alpha synthesis, coronary perfusion pressure (PP)
E A van Schaick et al.
Naunyn-Schmiedeberg's archives of pharmacology, 356(6), 827-837 (1998-02-07)
In this study we investigated the relationship between the pharmacokinetics and the cardiovascular and electroencephalogram (EEG) effects of three adenosine agonists with differing lipophilicity. Conscious normotensive rats received either 600 microg/kg N6-(p-sulphophenyl) adenosine (SPA), 200 microg/kg N6-cyclopentyladenosine (CPA) or 600
M S Dar
Pharmacology, biochemistry, and behavior, 37(4), 747-753 (1990-12-01)
To further investigate if the modulation of ethanol-induced motor incoordination is by brain adenosine A1 and/or A2 receptor, adenosine analogs with wide variability in their affinity for A1 and A2 subtypes were administered ICV and their effect on ethanol-induced (IP)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service