Skip to Content
Merck
All Photos(1)

Key Documents

C7740

Sigma-Aldrich

Anti-phospho-α-B Crystallin (14-25) (pSer19) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, bovine, human, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10 μg/mL
indirect immunofluorescence: 5 μg/mL
western blot: 0.5 μg/mL using human cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer19)

Gene Information

human ... CRYAB(1410)
mouse ... Cryab(12955)
rat ... Cryab(25420)

Specificity

Does not detect the unphosphorylated protein.

Immunogen

synthetic phosphopeptide surrounding the region of human α-B crystallin that contains serine 19.

Application

Anti-phospho-α-B Crystallin (14-25) (pSer19) antibody produced in rabbit
  • Immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 10μg/mL
  • Indirect immunofluorescence at a concentration of 5μg/mL
  • Western blotting at a concentration of 0.5μg/mL using human cell extract
  • It also used in 1- and 2-D immune blotting analysis.

Biochem/physiol Actions

α-Crystallin B chain is a protein encoded by the CRYAB gene in humans. αB-crystallin (ABC) and HSP27 are most abundant in human tissues. It is predominantly expressed in triple-negative breast cancer (TNBC) and leads to aggressive tumor phenotype. αB-crystallin expression is primary found in breast carcinomas and brain metastases. αB-crystallin is overexpressed or silenced in two different TNBC cell lines. It is a novel regulator of brain metastasis in TNBC and acts as a potential biomarker for drug target in aggressive disease. αB-crystallin and VEGF expressions in human conjunctival squamous cell carcinoma (CSCCs) may play an important role in the pathogenesis. Mutation in the gene is responsible for a number of inherited human disorders, which include cardiomyopathy, skeletal muscle myopathy and cataracts.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 3% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Ueda et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Protein homeostasis, including protein folding, refolding, and degradation, is thought to decline with aging. HSPB5 (also known as αB-crystallin) prevents target protein aggregation as a molecular chaperone and exhibits a cytoprotective function against various cell stresses. To elucidate the effect
Zhenyu Dong et al.
Anticancer research, 33(9), 3745-3751 (2013-09-12)
To examine the expression of αB-crystallin and vascular endothelial growth factor (VEGF) in conjunctival squamous cell carcinoma (CSCC). Seven CSCCs and three normal conjunctivas that were surgically excised were studied. Paraformaldehyde-fixed, paraffin-embedded tissue sections were processed for immunohistochemistry with antibodies
Bimal Prasanna Mohanty et al.
Indian journal of biochemistry & biophysics, 48(1), 35-41 (2011-04-08)
Crystallins are a diverse group of proteins that constitute nearly 90% of the total soluble proteins of the vertebrate eye lens and these tightly packed crystallins are responsible for transparency of the lens. These proteins have been studied in different
Dmitry Malin et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(1), 56-67 (2013-10-18)
Basal-like breast tumors are typically (ER/PR/HER2) triple-negative and are associated with a high incidence of brain metastases and poor clinical outcomes. The molecular chaperone αB-crystallin is predominantly expressed in triple-negative breast cancer (TNBC) and contributes to an aggressive tumor phenotype
Heng B Xie et al.
PLoS genetics, 9(6), e1003544-e1003544 (2013-07-03)
Dominant mutations in the alpha-B crystallin (CryAB) gene are responsible for a number of inherited human disorders, including cardiomyopathy, skeletal muscle myopathy, and cataracts. The cellular mechanisms of disease pathology for these disorders are not well understood. Among recent advances

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service