Skip to Content
Merck

C7002

Z-Val-Phe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H26N2O5
CAS Number:
Molecular Weight:
398.45
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

O=C(N[C@@H](C(C)C)C(N[C@H](C(O)=O)CC1=CC=CC=C1)=O)OCC2=CC=CC=C2

InChI

1S/C22H26N2O5/c1-15(2)19(24-22(28)29-14-17-11-7-4-8-12-17)20(25)23-18(21(26)27)13-16-9-5-3-6-10-16/h3-12,15,18-19H,13-14H2,1-2H3,(H,23,25)(H,24,28)(H,26,27)

InChI key

XINBRUNUJFZFGH-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



D Sindram et al.
Transplantation, 68(1), 136-140 (1999-07-31)
Cold preservation of the liver followed by reperfusion results in sinusoidal endothelial cell (SEC) apoptosis. Calpain-like activity is dramatically increased during reperfusion and inhibition of calpains results in lower graft injury and longer survival. Recently, calpains have been implicated in
V Kohli et al.
Gastroenterology, 116(1), 168-178 (1998-12-31)
Calpain proteases have been implicated in cell death by necrosis and more recently by apoptosis. Experiments were designed to determine the role of calpain proteases in ischemic rat liver injury by measurement of cytosolic calpain activity after different periods of
Heike Weber et al.
International journal of experimental pathology, 90(4), 387-399 (2009-08-08)
The cytosolic cysteine protease calpain is implicated in a multitude of cellular functions but also plays a role in cell damage. Our previous results suggest that an activation of calpain accompanied by a decrease in its endogenous inhibitor calpastatin may