Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C9H10ClNO3 · HCl
CAS Number:
Molecular Weight:
252.09
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
assay
≥95%
storage temp.
−20°C
SMILES string
Cl.NC(Cc1ccc(O)c(Cl)c1)C(O)=O
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J J Provencio et al.
Neurocritical care, 12(2), 244-251 (2009-12-08)
Cerebral vasospasm is a significant cause of morbidity in patients after aneurysmal subarachnoid hemorrhage (aSAH). There are few effective treatments. The search for new treatments has focused predominantly on dilating cerebral blood vessels. Growing evidence supports a role for inflammation
Laetitia Mouls et al.
Proteomics, 9(6), 1617-1631 (2009-03-03)
Protein oxidation is thought to contribute to a number of inflammatory diseases, hence the development of sensitive and specific analytical techniques to detect oxidative PTMs (oxPTMs) in biological samples is highly desirable. Precursor ion scanning for fragment ions of oxidized
Eline Thomson et al.
Free radical biology & medicine, 49(9), 1354-1360 (2010-07-22)
We aimed to determine whether myeloperoxidase (MPO) is the main peroxidase present in the airways of children with cystic fibrosis (CF) and to assess which oxidants it produces and whether they are associated with clinical features of CF. Children with
Charles O'Donnell et al.
COPD, 7(6), 411-417 (2010-12-21)
The airways of patients with chronic obstructive pulmonary disease (COPD) are associated with increased numbers of inflammatory cells, in particular neutrophils and macrophages. Contained within the primary granules of neutrophils is the heme enzyme called myeloperoxidase (MPO) that has been
Michael Holzer et al.
Antioxidants & redox signaling, 14(12), 2337-2346 (2011-01-18)
Carbamylation of proteins through reactive cyanate has been demonstrated to predict an increased cardiovascular risk. Cyanate is formed in vivo by breakdown of urea and at sites of inflammation by the phagocyte protein myeloperoxidase. Because myeloperoxidase (MPO) associates with high-density
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service