Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 ea | Please contact Customer Service for Availability | CZK 2,540.00 |
100 ea | Please contact Customer Service for Availability | CZK 7,580.00 |
About This Item
NACRES:
NA.56
UNSPSC Code:
41105500
CZK 2,540.00
Please contact Customer Service for Availability
form
tablet
Quality Segment
usage
tablet sufficient for 1 mL lysis (cell suspension)
storage temp.
−20°C
Application
All the advantages of CelLytic™ Express now in a tablet format. For direct lysis of bacterial cultures and for use in the His-Select® iLAP™ column. CelLytic Express tablets are a complete formulation including lysozyme and endonuclease for complete cell lysis. The resulting lysate is clear of cellular debris, and is immediately ready for standard affinity purification. This new formulation adds minimal volume to the final lysate, providing a method with defined volume and time parameters, making it easier to validate and making it more convenient for production scale protocols.
Features and Benefits
- Efficient and non-denaturing in-culture protein extraction
- Save time by eliminating cell harvest and clarification steps
- Preserve biological activity with fewer manipulations
- Compatible with affinity resins and magnetic beads
- Ideal for Production Scale Extraction protocols
Other Notes
One unit will lyse 0.6 μg of Micrococcus lysodeikticus per minute by turbidimetric detection at 600 nm when suspended in buffer at pH 6.2 at 25 °C.
Legal Information
CelLytic is a trademark of Sigma-Aldrich Co. LLC
HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany
iLAP is a trademark of Sigma-Aldrich Co. LLC
1 of 1
This Item | |||
|---|---|---|---|
| usage tablet sufficient for 1 mL lysis (cell suspension) | usage - | usage - | usage 1 kit sufficient for 5 g fresh or frozen cell paste lysis. |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. room temp | storage temp. −20°C |
| form tablet | form powder | form solution | form - |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




