Skip to Content

C5384

Cholesteryl heptadecanoate

Synonym(s):

3β-Hydroxy-5-cholestene 3-heptadecanoate, 5-Cholesten-3β-ol 3-heptadecanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H78O2
CAS Number:
Molecular Weight:
639.09
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95%

SMILES string

CCCCCCCCCCCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C44H78O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-24-42(45)46-37-29-31-43(5)36(33-37)25-26-38-40-28-27-39(35(4)23-21-22-34(2)3)44(40,6)32-30-41(38)43/h25,34-35,37-41H,7-24,26-33H2,1-6H3/t35-,37+,38+,39-,40+,41+,43+,44-/m1/s1

InChI key

PPQNZVDOBYGOLY-BFGJSWSOSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library