Skip to Content
Merck

Skip To

C-41110

Trypsin Neutralizing Solution (TNS)

0.05% Trypsin Inhibitor in 0.1% BSA, 125 ml

Synonym(s):

Neutralizing Solution, TNS Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
125 mL
Please contact Customer Service for Availability
3 110,00 CZK

About This Item

NACRES:
NA.81
UNSPSC Code:
12352207

3 110,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

packaging

pkg of 125 ml

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Trypsin Neutralizing Solution (TNS) (0.05% Trypsin Inhibitor in 0.1% BSA)

Application

The Trypsin Neutralizing Solution contains 0.05% Trypsin Inhibitor in 0.1% BSA and is available in different sizes.

Preparation Note

Click here for more information.
Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label.

Analysis Note

All lots of PromoCell Cell Detachment Reagents are subjected to comprehensive quality control tests. Each lot is routinely tested for biological function, absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters