Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
125 mL | Please contact Customer Service for Availability | 1 630,00 CZK |
About This Item
NACRES:
NA.78
UNSPSC Code:
12352207
form
aqueous solution
packaging
pkg of 125 ml
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Trypsin / EDTA (0.04% / 0.03%)
Application
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This Trypsin/EDTA Solution has a ratio of 0.04% trypsin and 0.03% EDTA and is available in different sizes.
Preparation Note
Click here for more information.
Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label.
Analysis Note
All lots of PromoCell Cell Detachment Reagents are subjected to comprehensive quality control tests. Each lot is routinely tested for biological function, absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| form aqueous solution | form aqueous solution | form aqueous solution | form solution |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| packaging pkg of 125 ml | packaging pkg of 30 ml | packaging pkg of 250 ml | packaging - |
Still not finding the right product?
Explore all of our products under Trypsin / EDTA
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



