Skip to Content
Merck

C3242

CDKi hydrochloride

≥98% (HPLC), solid

Synonym(s):

2,4-Diamino-5-oxo-pyrimidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H6N4O · xHCl
CAS Number:
Molecular Weight:
126.12 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

solubility

H2O: 21 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].Nc1ncc(O)c(N)n1

InChI

1S/C4H6N4O.ClH/c5-3-2(9)1-7-4(6)8-3;/h1,9H,(H4,5,6,7,8);1H

InChI key

KEFSYBXFZWLDQU-UHFFFAOYSA-N

Biochem/physiol Actions

CDKi hydrochloride is a potent and selective ATP competitiive inhibitor of cycB-CDK1, cycE-CDK2, and cycD1-CDK4. Demonstrates significant inhibitory activity against a variety of tumor type both in vitro and in vivo.

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library