Merck
Skip To
C2798
Biohazard incinerator carton
Floor model
Synonym(s):
incineration waste container, 132050001, F13205-0001, lab waste carton, waste canister, waste receptable
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
6 ea
Please contact Customer Service for Availability
CZK 4,010.00
About This Item
NACRES:
NB.35
UNSPSC Code:
41100000
CZK 4,010.00
Please contact Customer Service for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Bel-Art F13205-0001
size
12 in. × 12 in. × 27 in.
General description
Designed to be incinerated along with its contents when full for safer disposal of biohazardous material. The sturdy corrugated cardboard box is supplied with a 1.5 mil HMHD liner to contain liquid waste and is imprinted with the "biohazard" pictogram and instructions in four languages.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
BAF132050002TMO6370-0004Z370665
size
12 in. × 12 in. × 27 in.
size
-
size
-
size
-
manufacturer/tradename
Bel-Art F13205-0001
manufacturer/tradename
Bel-Art F13205-0002
manufacturer/tradename
Nalgene 6370-0004
manufacturer/tradename
Nalgene 6370-0005
Choose from one of the most recent versions:
Certificates of Analysis (COA)