Skip to Content
Merck
All Photos(1)

Key Documents

C2460

Sigma-Aldrich

Casein Kinase II substrate

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H83N21O28
CAS Number:
Molecular Weight:
1450.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

potency

19 μM Km

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C52H83N21O28/c1-19(63-40(91)21(6-3-9-61-51(56)57)65-41(92)22(7-4-10-62-52(58)59)64-39(90)20(53)5-2-8-60-50(54)55)38(89)66-23(11-31(75)76)42(93)70-28(16-36(85)86)47(98)73-30(18-74)48(99)71-26(14-34(81)82)45(96)68-24(12-32(77)78)43(94)67-25(13-33(79)80)44(95)69-27(15-35(83)84)46(97)72-29(49(100)101)17-37(87)88/h19-30,74H,2-18,53H2,1H3,(H,63,91)(H,64,90)(H,65,92)(H,66,89)(H,67,94)(H,68,96)(H,69,95)(H,70,93)(H,71,99)(H,72,97)(H,73,98)(H,75,76)(H,77,78)(H,79,80)(H,81,82)(H,83,84)(H,85,86)(H,87,88)(H,100,101)(H4,54,55,60)(H4,56,57,61)(H4,58,59,62)/t19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-/m0/s1

InChI key

NSTKIPFKPLZQKH-UPPQRMANSA-N

Amino Acid Sequence

Arg-Arg-Arg-Ala-Asp-Asp-Ser-Asp-Asp-Asp-Asp-Asp

Biochem/physiol Actions

Selective substrate for Casein Kinase II.

Substrates

This peptide cannot be phosphorylated by casein kinase-1.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Marin et al.
Biochemical and biophysical research communications, 198(3), 898-905 (1994-02-15)
The available information about the specificity determinants of casein kinases-1 and -2 (CK1 and CK2) has been utilized to obtain two new peptide substrates optimally suited for the specific monitoring of these two pleiotropic enzymes. The best substrate developed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service