Skip to Content
Merck

C2285

Cytochrome f from spinach

essentially salt-free, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

essentially salt-free, lyophilized powder

absorbance ratio

A554/280 nm ~0.5

storage temp.

−20°C

Preparation Note

Prepared by a modification of the method of Ho and Krogman, J. Biol. Chem., 255, 3855 (1980).

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service