Skip to Content
Merck

Skip To

C2247

CPPHA

≥98% (HPLC)

Synonym(s):

N-[4-Chloro-2-(phthalimidomethyl)phenyl]salicylamide, N-[4-Chloro-2-[(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]phenyl}-2-hydroxybenzamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H15ClN2O4
CAS Number:
Molecular Weight:
406.82
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated


assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: >20 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

−20°C

SMILES string

Oc1ccccc1C(=O)Nc2ccc(Cl)cc2CN3C(=O)c4ccccc4C3=O

InChI

1S/C22H15ClN2O4/c23-14-9-10-18(24-20(27)17-7-3-4-8-19(17)26)13(11-14)12-25-21(28)15-5-1-2-6-16(15)22(25)29/h1-11,26H,12H2,(H,24,27)

InChI key

UFOUABRZSDGGAZ-UHFFFAOYSA-N

Biochem/physiol Actions

CPPHA is a positive allosteric modulator of mGluR5 receptor.
CPPHA is a selective positive allosteric modulator of mGluR5 receptor. It has no agonist activity alone, but reduces threshold response and shifts dose-response curves to glutamate, quisqualate, and DHPG by 4- to 7-fold to the left in recombinant CHO cells expressing human or rat mGluR5.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters