Skip to Content
Merck

Skip To

C-12975

Human Mesenchymal Stem Cells (hMSC)

Derived from bone marrow, 500,000 proliferating cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
39 500,00 CZK

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human bone marrow
Morphology:
(stromal)

39 500,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human bone marrow

packaging

pkg of 500,000 cells

morphology

(stromal)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Application

Mesenchymal Stem Cells (MSC), also termed Mesenchymal Stromal Cells, are multipotent cells that can differentiate into a variety of cell types and have the capacity for self renewal. MSC have been shown to differentiate in vitro or in vivo into adipocytes, chondrocytes, osteoblasts, myocytes, neurons, hepatocytes, and pancreatic islet cells. Optimized PromoCell media are available to support both the growth of MSC and their differentiation into several different lineages. Recent experiments suggest that differentiation capabilities into diverse cell types vary between MSC of different origin.Human Mesenchymal Stem Cells (hMSC-BM) are harvested from normal human bone marrow of individual donors and are provided in a proliferating format. The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, and osteoblasts. The cells show a verified marker expression profile that complies with ISCT recommendations, providing well characterized cells (Cytotherapy (2006) Vol. 8, No. 4, 315-317).

Biochem/physiol Actions

Bone Marrow

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of Mesenchymal Stem Cells. They are tested for cell morphology, proliferation potential, adherence rate, and viability. Furthermore, they are characterized by flow cytometric analysis of a comprehensive panel of markers, namely CD73/CD90/CD105 and CD14/CD19/CD34/CD45/HLA-DR as proposed by the ISCT. Differentiation assays into adipogenic, osteogenic and chrondrogenic are performed for each lot under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

Recommended Plating Density: 4000 cells per cm2Tested Markers: CD105 positive, CD73 positive, CD90 positive, CD45 negative, CD34 negative, CD14 negative, CD19 negative, HLA-DR negativeDifferentiation capacity to adipogenic, osteogenic and chondrogenic cells
Recommended Primary Cell Culture Media:Link
Recommended Primary Cell Culture Media:Link

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C12971C14091C12972
description

Derived from bone marrow, 500,000 proliferating cells

description

Derived from umbilical cord matrix, 500,000 cryopreserved cells

description

Derived from umbilical cord matrix, 1 million cells per pellet

description

Derived from umbilical cord matrix, 500,000 proliferating cells

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

biological source

human bone marrow

biological source

human umbilical cord matrix (Wharton′s jelly)

biological source

human umbilical cord matrix (Wharton′s jelly)

biological source

human umbilical cord matrix (Wharton′s jelly)

morphology

(stromal)

morphology

(stromal)

morphology

(stromal)

morphology

(stromal)

shipped in

ambient

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

storage temp.

room temp

storage temp.

−196°C

storage temp.

−20°C

storage temp.

room temp

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 1,000,000 cells

packaging

pkg of 500,000 cells


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

10108-64-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters