Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human (bronchiole)
Morphology:
(epithelial)
Relevant disease(s):
asthma
biological source
human (bronchiole)
packaging
pkg of 500,000 cells
morphology
(epithelial)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
asthma
shipped in
ambient
storage temp.
room temp
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Application
Primary Human Small Airway Epithelial Cells (HSAEpC) are isolated from the distal portion of the human respiratory tract in the 1 mm bronchiole area and are provided in a proliferating format. They stain positive for cytokeratin.The distal respiratory tract mainly consists of pulmonary alveoli, which are spherical outcroppings of the respiratory bronchioles and are the primary sites of gas exchange with the blood. HSAEpC are qualified for functional studies to investigate disorders such as asthma or pulmonary inflammation.
Biochem/physiol Actions
Bronchiole
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of Epithelial Cells. They are tested for cell morphology, adherence rate and cell viability. Furthermore, flow cytometric analyses for the cell-type specific marker cytokeratin are carried out for each lot. Growth performance is tested through multiple passages under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
Recommended Plating Density: 10000 - 15000 cells per cm2Passage After Thawing: P2Tested Markers: Cytokeratin positiveGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Still not finding the right product?
Explore all of our products under Human Small Airway Epithelial Cells (HSAEpC)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7791-20-0
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.