Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human heart (ventricles)
Morphology:
(fibroblast)
Relevant disease(s):
cardiovascular diseases
biological source
human heart (ventricles)
packaging
pkg of 500,000 cells
morphology
(fibroblast)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cardiovascular diseases
shipped in
ambient
storage temp.
room temp
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Application
Primary Human Cardiac Fibroblasts (HCF) are isolated from the ventricles of the adult heart and are provided in a proliferating format. They play a central role in the maintenance of the extracellular matrix in the normal heart and the synthesis of growth factors and cytokines. Under pathophysiological conditions, cardiac fibroblasts are involved in scar formation after myocardial infarction, cardiac fibrosis, and cardiac hypertrophy. HCF can be used to study such processes in vitro.
Biochem/physiol Actions
Heart
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of Human Fibroblasts. They are tested for cell morphology, adherence rate and cell viability. Flow cytometric analyses of the fibroblast specific antigen CD90 are performed for each lot. Growth performance is tested through multiple passages up to 10 or 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
Recommended Plating Density: 3500 - 7000 cells per cm2Passage After Thawing: P2Tested Markers: CD90 positive, Smooth muscle alpha-actin negative, slow muscle myosin negativeGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Still not finding the right product?
Explore all of our products under Human Cardiac Fibroblasts (HCF)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.