Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
O[Ge(=O)CH2CH2CO2H]2
CAS Number:
Molecular Weight:
339.42
UNSPSC Code:
12352103
PubChem Substance ID:
EC Number:
235-800-0
MDL number:
Pricing and availability is not currently available.
SMILES string
OC(=O)CC[Ge](=O)O[Ge](=O)CCC(O)=O
InChI
1S/C6H10Ge2O7/c9-5(10)1-3-7(13)15-8(14)4-2-6(11)12/h1-4H2,(H,9,10)(H,11,12)
InChI key
XEABSBMNTNXEJM-UHFFFAOYSA-N
Biochem/physiol Actions
An organic germanium-containing compound that induces natural killer T cells in mice. These cells secrete interferon-γ. IFN-γ levels were elevated 20 hours after administration of 300 mg/kg.
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.