Skip to Content
Merck

C0480

Z-Asp(OMe)-Gln-Met-Asp(OMe) fluoromethyl ketone

powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C29H40N5O11FS
CAS Number:
Molecular Weight:
685.72
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

assay

≥90% (TLC)

form

powder

color

white

solubility

acetone: absolute methanol (1: 1): 1 mg/mL, clear, colorless (
acetone:Absolute methanol(1:1))

shipped in

dry ice

storage temp.

−20°C

SMILES string

COC(=O)C[C@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(=O)OC)NC(=O)OCc1ccccc1)C(=O)CF

InChI

1S/C29H40FN5O11S/c1-44-24(38)13-20(22(36)15-30)34-27(41)19(11-12-47-3)33-26(40)18(9-10-23(31)37)32-28(42)21(14-25(39)45-2)35-29(43)46-16-17-7-5-4-6-8-17/h4-8,18-21H,9-16H2,1-3H3,(H2,31,37)(H,32,42)(H,33,40)(H,34,41)(H,35,43)/t18-,19-,20-,21-/m0/s1

InChI key

QFITYVNVMNJELE-TUFLPTIASA-N

Biochem/physiol Actions

A cell-permeable inhibitor of caspase 3 and 6, which exhibits competitive and irreversible inhibition.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library