Skip to Content
Merck

BR701376

BRAND® sealing film for microplates

suitable for PCR, storage, PET film, DMSO/solvent resistant, operating temperature (-80)—120 °C, pack of 100 ea

Synonym(s):

cover, plate film, seal, sealing film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122410
NACRES:
NB.43
eCl@ss:
32120613
Manufacturer/tradename:
BRAND 701376
Material:
PET film
Sterility:
non-sterile
Packaging:
pack of 100 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

PET film

sterility

non-sterile

feature

DNA free, RNase and DNase free

packaging

pack of 100 ea

manufacturer/tradename

BRAND 701376

parameter

(-80)—120 °C operating temperature

suitability

suitable for PCR

General description

This film is ideal for PCR, ELISA and sample storage. Due to its adhesive, it can be used down to -80°C, is DMSO-resistant, and resistant to common solvents. It can be removed without leaving any residue so that work can continue unhindered after transport or storage.

Features and Benefits

  • Temperature stability to 120°C and down to -80°C
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant
  • Removable without leaving residue

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library