Skip to Content
Merck

BM0028

BMS-536924

≥98% (HPLC)

Synonym(s):

4-[[(2S)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]-3-[7-methyl-5-(4-morpholinyl)-1H-benzimidazol-2-yl]-2(1H)-pyridinone, BMS 536924

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H26ClN5O3
CAS Number:
Molecular Weight:
479.96
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CC(N2CCOCC2)=CC3=C1NC(C4=C(NC[C@@H](O)C5=CC(Cl)=CC=C5)C=CNC4=O)=N3

Biochem/physiol Actions

BMS-536924 is a potent, orally active, ATP-competitive insulin-like growth gactor-1 receptor (IGF1R) inhibitor.
BMS-536924 is a potent, orally active, ATP-competitive insulin-like growth gactor-1 receptor (IGF1R) inhibitor that shows anticancer activities in preclinical models.

Features and Benefits

BMS-536924 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library