Skip to Content
Merck

BCS-406

CoolCell 5 mL LX Cell Freezing Container

for 12 x 3.5 mL to 5 mL cryogenic vials, purple

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
41103025
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

purple

feature

capacity 12 tubes, for tubes 3.5 ‑ 5 mL

manufacturer/tradename

BioCision BCS-406

diam. × H

9.5 × 14.5 cm

well diam.

15.2 mm

General description

CoolCell 5ml LX alcohol-free freezing container holds 12, 3.5 mL to 5.0 mL cryogenic vials and provides a highly reproducible -1 degree C/minute freezing rate when used in a -80 degree C freezer. No alcohol or fluids required. The insulative outer housing and thermo-conductive core ensure consistent heat removal to all vials throughout the freezing period. CoolCell containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen - no "frosty" fingers. Enhanced design features include beveled junction between base and lid to allow quicker opening once frozen, numbered vial wells for easy sample identification, and exposed vial tops when container is open allowing for faster removal of vials for placement onto dry ice or liquid nitrogen.

Features and Benefits

Save money and go green! If used regularly, CoolCell containers save approximately $350/year per unit over isopropanol-based freezing containers by eliminating alcohol purchase and hazardous waste disposal.

Holds 12 x 3.5 mL to 5.0 mL cryogenic vials. Cryogenic vials fill volume: 3.5 mL to 5.0 mL per vial. Fill empty wells with CoolCell Filler Vials, 5 mL to ensure constant -1 degree C/minute freeze rate.

Legal Information

CoolCell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service